Imprimer
Solid phase extraction cartridges, HyperSep™ Aminopropyl
Fournisseur: Thermo Fisher Scientific
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
60108-519 60108-432 60108-518 60108-364 60108-424 60108-425 60108-738 60108-739 60108-740
HYPE60108-519EA 0 EUR
HYPE60108-519 HYPE60108-432 HYPE60108-518 HYPE60108-364 HYPE60108-424 HYPE60108-425 HYPE60108-738 HYPE60108-739 HYPE60108-740
Solid phase extraction cartridges, HyperSep™ Aminopropyl
Colonnes pour chromatographie
A polar sorbent for both polar and anion exchange interactions.
  • Weak anion exchanger
  • Enhanced retention of strong anions, such as sulfonic acids
  • Excellent retention of drugs, metabolites and structural isomers