Imprimer
Screw caps for screw neck vials, N 13
Fournisseur: MACHEREY-NAGEL
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
702926 702963 702966 702052 702050 702051 702113
MANA702926EA 46.4 EUR
MANA702926 MANA702963 MANA702966 MANA702052 MANA702050 MANA702051 MANA702113
Screw caps for screw neck vials, N 13
Capuchons Bouchons pour flacons de chromatographie
The screw caps with assembled septa are ready-to-use.
  • Caps with center hole as well as with closed top
  • Bonded screw cap (septa firmly connected with the cap) for blunt HPLC needles is available
  • Septa made from economical Red Rubber/FEP and high quality silicone/PTFE
  • Plain caps and an assortment of single septa including virgin PTFE septa