Imprimer
Disposable earplugs, 304
Fournisseur: HONEYWELL SÉCURITÉ
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
1000106 1000107
111-1335EA 51.31 EUR
111-1335 111-1336
Disposable earplugs, 304
Earplugs Disposable Earplugs
These striped PVC-free foam plugs are available in two sizes to ensure a proper and comfortable fit.
  • PU foam expands gently in the ear canal, reduces tendency to back out of the ear canal
  • Smooth, soil resistant skin prevents dirt build-up, promotes proper hygiene
  • Tapered bullet-shaped design easier to roll-down and insert
Suitable for medium/high noise work situations that demand a high degree of comfort, frequent changes, or where hygiene presents a problem for re-use.
SNR: 33 dB.
Certifications: EN 352-2:2002