Imprimer
Analytical balances, Quintix®
Fournisseur: Sartorius Balances
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
QUINTIX125D-1S QUINTIX35-1S QUINTIX65-1S QUINTIX125D-EU
611-3290EA 7530 EUR
611-3290 611-3291 611-3292 611-3865
Analytical balances, Quintix®
Balances and Scales
A series of ergonomic balances designed for easy, intuitive operation, with a large touch screen, self explanatory icons and plain text prompts. In addition to the standard applications including general weighing, density, percentage, check weighing, peak hold, counting, and averaging, the Quintix® range also provides mixing, components, statistics and conversion funtionalities. Results can be transferred via a mini-USB interface to a printer or a computer directly into a Microsoft Excel® spreadsheet without additional software. Menu with choice of 10 languages.
  • High contrast colour touch screen
  • Stabilisation times for 4 place balances of 2-3 seconds with excellent repeatability (0,1 mg) and linearity (0,2 mg)
  • IsoCAL internal calibration and adjustment function
  • Draught shield with doors that glide smoothly open that also can be completely disassembled for easy cleaning
  • Supervisor lock to prevent any unintentional changes to settings
  • Optional GLP compliant YDP30 printer that is automatically detected by the Quintix® balance
Semi-micro balances, five-place: The introduction of five-place balances to the Quintix® series extends the level of performance of these compact, accurate balances.
Additional features include:
One touch draft shield door operation
Stabilisation times for five-place semi-micro balances are approximately 6 seconds
Start Assistant function and SmartTouch user interface to help guide the user through the range of functionalities available
Optional 90 mm weighing pan
The introduction of 5-place balances to the Quintix® series extends the level of performance to these compact accurate balances.
Additional features include:
Commander maintenant