Imprimer
Powder triers
Fournisseur: Sampling Systems
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
1216A-1500 1216A-600 1216A-1000
SAMP1216A-1500EA 2890 EUR
SAMP1216A-1500 SAMP1216A-600 SAMP1216A-1000
Powder triers
Échantillonneurs Préleveurs de solides
These triers are suitable for taking a large volume, cross sectional sample from a container.
  • Robust all stainless steel construction with bright finish
  • Cutting scroll for the sampler to go into the product
  • Turn the handle to close the slot
Suitable for sampling cereals, grains, free flowing powders, granules and even slightly cohesive or compacted powders.
Certifications: GMP
Commander maintenant
SPÉCIFICATIONS
Environmentally Preferable