Imprimer
Analytical balances, Secura®
Fournisseur: Sartorius Balances
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
SECURA125-1S SECURA225D-1S SECURA225D-CEU SECURA125-1CEU SECURA125-1CFR
611-3324EA 8390 EUR
611-3324 611-3325 SARBSECURA225D-CEU SARBSECURA125-1CEU SARBSECURA125-1CFR
Analytical balances, Secura®
Balances and Scales
Advanced balances designed for easy, reliable and secure weighing operations for regulated areas. These analytical balances deliver stabilisation times of 2-3 seconds with excellent repeatability (0,1 mg) and linearity (0,2 mg). The Secura® range features a range of APC functions (Advanced Pharma Compliance) to help with documentation and monitoring requirements. The GLP compliant printout documentation is also controlled by the inbuilt security control, as data transfer to a printer or PC is blocked, when common operating errors occur, such as the balance is not level, minimum sample weight has not been achieved or temperature fluctuations require isoCAL calibration adjustment.
  • Touch screen with graphical user interface optimised for users in pharmaceutical laboratories
  • Cal Audit Trail function to document the quality of each weigh-in procedure you perform
  • IsoCAL internal calibration and adjustment function linked to CAL Audit Trail
  • SQmin - monitoring of compliance with USP minimum sample weight limits
  • Password protection of set-up settings
  • LevelControl continuously checks to see if the balance is perfectly level
Semi-micro balances, five-place: The introduction of five-place balances to the Secura® series extends the level of performance to these compact accurate balances.
Additional features include:
One touch draft shield door operation
Stabilisation times for 5-place semi-micro balances are approximately 6 seconds
Start Assistant function and SmartTouch user interface to help guide the user through the range of functionalities available
Optional 90 mm weighing pan
Informations de livraison: Supplied with universal power adapter, in-use cover, application guide, dust cover for analytical balances and draught ring for 0,1 mg balances.