Imprimer
Caps for sample tubes, tamper evident
Fournisseur: Simport Scientific
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
T550W T550WOS T552-7WTP T552WTP T552-30WTP T552-30W T552W
720-0574EA 85.8 EUR
720-0574 216-1007 SIMPT552-7WTP SIMPT552WTP SIMPT552-30WTP SIMPT552-30W SIMPT552W
Caps for sample tubes, tamper evident
Capuchons Bouchons pour tubes à essais
Caps made of PE feature a long skirt and a super fast thread design allowing them to be removed or sealed with a single turn. Caps are designed with flexible sealing lip which provides leakproof seal.
Attention: *Le(s) produit(s) sur cette page sont hors du champ d'application de la directive européenne 2019/904