Imprimer
Sealing film for biological cultures
Fournisseur: VWR Collection
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
731-0315EA 150 EUR
731-0315 731-0316 731-0313 731-0314
Sealing film for biological cultures
Seals Microplate Seals
Rayon film with medical grade adhesive. Hydrophobic, porous film in sheeted roll format for sealing tissue culture plates, bio-blocks, and 96-well plates where gas exchange is necessary for cell or bacterial growth.
  • Recommended temperature range from −20 to +80 °C
  • Total thickness 139,7 μm
  • Pre-scored films separated by red colour bands for easy application
  • Pierceable and breathable
  • Pre-cut films on a roll
Informations de livraison: Each roll contains sufficient film to cover 50 plates.