Imprimer
Safety ankle boots, lace-up, Jalterre
Fournisseur: Jallatte
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
JJV31/38 JJV31/39 JJV31/40 JJV31/41 JJV31/42 JJV31/43 JJV31/44 JJV31/45 JJV31/46 JJV31/47
JALCJJV31/38EA 269 EUR
JALCJJV31/38 JALCJJV31/39 JALCJJV31/40 JALCJJV31/41 JALCJJV31/42 JALCJJV31/43 JALCJJV31/44 JALCJJV31/45 JALCJJV31/46 JALCJJV31/47
Safety ankle boots, lace-up, Jalterre
Chaussures et bottes
Anti-static ankle boots made from full grain water repellent leather upper, Gore-Tex® Performance lining and Kompact Vibram® sole (fuil and oil resistant; contact heat resistant up to 300 °C HRO); with padded tongue and ergonomic collar. Suitable for off-shore and construction.
  • Lining made from Gore-Tex® membrane
  • Confort-Tech integrated insole: anatomic, antibacterial, shock absorber in the heel area
  • Non magnetic, lightweight 200 J toecap (100% composite fibre); perforated for perfect foot ventilation
  • Non magnetic, lightweight and flexible FleXtane™ HP midsole (100% composite)
  • Compact Vibram® sole with superb adherence and grip, high anti-abrasion value and 6,5 mm high self-cleaning cleats
Colour: Black
Certifications: EN ISO 20345:2007, S3, CI, WR, HRO, SRC