Imprimer
Single channel micropipettes, electronic, variable volume, Acura® electro 926
Fournisseur: Socorex Isba
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
926.1000E 926.0100E 926.0200 926.0010 926.0200E 926.1000 926.0002E 926.0002G 926.0010E 926.0010G 926.0010Y 926.0010YE 926.0010YG 926.0020 926.0020E 926.0020G 926.0050 926.0200G 926.1000G 926.0002 926.0100 926.0100G 926.0050G 926.0050E
SOCO926.1000EEA 1420 EUR
SOCO926.1000E SOCO926.0100E SOCO926.0200 SOCO926.0010 SOCO926.0200E SOCO926.1000 SOCO926.0002E SOCO926.0002G SOCO926.0010E SOCO926.0010G SOCO926.0010Y SOCO926.0010YE SOCO926.0010YG SOCO926.0020 SOCO926.0020E SOCO926.0020G SOCO926.0050 SOCO926.0200G SOCO926.1000G SOCO926.0002 SOCO926.0100 SOCO926.0100G SOCO926.0050G SOCO926.0050E
Single channel micropipettes, electronic, variable volume, Acura® electro 926
Pipettes Pipetteurs
These micropipettes include a microprocessor-controlled motor and a rechargeable battery pack.
  • Reduced shaft length and conical end improve driveability
  • Easy access to microtubes and microplates
  • Unsurpassed metrological performance and stability
  • Interchangeable volumetric modules available separately: Disassembling and re-assembling without any tool contributes towards maximal flexibility
Informations de commande: The pipette packages are supplied with individual QC certificate and operating instructions, pipette charging stand, fast charger with AC adapter and cord, additional battery pack and Qualitips® pipette tip samples.
The individual pipettes are supplied with QC certificate and operating instructions, as well as samples of Qualitips®.