Imprimer
HATU (O-(7-Aza-1H-benzotriazol-1-yl)-N,N,N',N'-tetramethyluronium hexafluorophosphate), Novabiochem®
Marques : Novabiochem (Part of Merck)
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
Danger
Synonyme(s): HATU, 1-[Bis(dimethylamino)methylene]-1H-1,2,3-triazolo[4,5-b]pyridinium 3-oxide, hexafluorophosphate
8.51013.0025 8.51013.0100 8.51013.0500 8.51013.1000
01-62-0041-25EA 497 EUR
01-62-0041-25 8.51013.0100 NOVA8.51013.0500 NOVA8.51013.1000
HATU (O-(7-Aza-1H-benzotriazol-1-yl)-N,N,N',N'-tetramethyluronium hexafluorophosphate), Novabiochem®
HATU (O-(7-Aza-1H-benzotriazol-1-yl)-N,N,N',N'-tetramethyluronium hexafluorophosphate)
A highly efficient coupling reagent for solid and solution phase peptide synthesis.
In comparative studies HATU has been found to give better coupling yields with less enantiomerization that HBTU, TBTU or PyBOP® [1-3]. It is particularly effective at coupling to N-alkyl amines where other coupling reagents give poor yields and is the preferred reagent for loading resins bearing secondary amino groups.
Formule: C₁₀H₁₅F₆N₆OP
Poids moléculaire: 380.2 g/mol
Point de fusion: 186 °C
Température de stockage: Réfrigérateur
Numéro MDL: MFCD00274639
Numéro CAS: 148893-10-1