Imprimer
Bottles, wide neck, LDPE, coloured, with screw cap
Fournisseur: KJS KUNSTSTOFFVERPACKUNGEN
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
WM50RO WM500RO WM50BL WM250BL WM50GR WM1000GR WM1000GE
215-6031EA 20.5 EUR
215-6031 215-6034 215-6036 215-6038 215-6041 215-6045 215-6050
Bottles, wide neck, LDPE, coloured, with screw cap
Flacons Flacons en plastique
HDPE, coloured.
  • Good chemical resistance
  • Good temperature resistance