Imprimer
Wash bottles, narrow or wide neck
Fournisseur: VITLAB
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
94993 93793 95093 93993 95193 94193
VITL94993EA 7 EUR
VITL94993 VITL93793 VITL95093 VITL93993 VITL95193 VITL94193
Wash bottles, narrow or wide neck
Flacons Pissettes
PP, translucent, screw cap and spray tube made from PP.
  • Excellent chemical resistance
  • Excellent temperature resistance
Precise spray jet and optimised medium backflow through the continuous and finely drawn tip of the spray tube.