Imprimer
Handles for Glass Beakers
Fournisseur: BOCHEM
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
8993 8995 8996
BOCH8993EA 67.1 EUR
BOCH8993 BOCH8995 BOCH8996
Handles for Glass Beakers
Carriers Bottle Carriers/Containers
These handles are made of stainless steel 18/10 and are used to carry glass beakers.