Imprimer
Single channel micro pipettes, variable volume, Acura® manual 825
Fournisseur: Socorex Isba
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
825.0002 825.0010 825.0010Y 825.0020 825.0050 825.0100 825.0200 825.1000 825.TRIO.C 825.TRIO.J 825.TRIO.M 825.TRIO.P 825.TRIO.S 825.TRIO.V 825.TRIO.T 825.TRIO.O
613-4870EA 465 EUR
613-4870 613-4871 613-4872 613-4873 613-4874 613-4875 613-4876 613-4877 613-7246 613-7247 613-7248 613-7250 613-7251 SOCO825.TRIO.V 613-7252 613-7249
Single channel micro pipettes, variable volume, Acura® manual 825
Pipettes Pipetteurs
Manual pipettes for all applications
  • Single-handed volume setting
  • Precision digital display visible at all time
  • Justip™ adjustable tip ejector fitting most tips
Informations de livraison: The Multisets include: Three Acura® 825 manual adjustable pipettes, Qualitips® samples, QC certificates, operating instructions