Imprimer
HPLC columns, Syncronis™ Amino
Fournisseur: Thermo Fisher Scientific
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
97705-014001 97702-052130 97702-102130 97705-154630 97705-254630 97705-152130
HYPE97705-014001EA 0 EUR
HYPE97705-014001 HYPE97702-052130 HYPE97702-102130 HYPE97705-154630 HYPE97705-254630 HYPE97705-152130
HPLC columns, Syncronis™ Amino
Colonnes pour chromatographie
These columns are designed with a versatile aminopropyl phase that gives excellent chromatographic properties in four modes: weak anion exchange, reversed phase, normal phase and HILIC.
  • Highly pure, high surface area silica
  • Double endcapped for extra surface coverage
Surface area: 320 m²/g
Carbon load: 4%
pH range: 2 to 8