Imprimer
Plaques de filtration, MultiScreen
Fournisseur: Merck
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
MAHAN4550 MAHAS4510 MSHAN4510 MSHAS4510 MSHAN4550 MSHAN4B50 MSHAS4B10
736-0183EA 1450 EUR
736-0183 736-0184 MILFMSHAN4510 MILFMSHAS4510 MILFMSHAN4550 MILFMSHAN4B50 MILFMSHAS4B10
Plaques de filtration, MultiScreen
Microplaques Plaques de filtration
MultiScreenHTS plates are specifically developed for high-throughput use with automated work stations. Rigid sidewalls improve handling and ample surfaces are provided for bar code labels.
  • Hydrophilic mixed cellulose esters (MCE) membrane
Wells are individually sealed to prevent incubation crosstalk and the underdrain is removable for access to the filters.
MultiScreen HTS 96-well filtration plates are used to filter samples and perform entire procedures, from cell growth to scintillation counting within the same plate. The plates come in many membrane types, pore sizes, and plate materials. The automation-compatible design allows for easy manipulation with a variety of robotics systems and makes barcoding possible on all four sides. Filtration is accomplished either by vacuum using the MultiScreen HTS Vacuum Manifold, or by centrifugation.