Imprimer
Spider evaporators
Fournisseur: BUCHI
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
001332 001333 001334 001335 001336 011574 011575 011576 011577 011578
531-1319EA 1870 EUR
531-1319 531-1320 531-1321 531-1322 531-1323 BUCH011574 BUCH011575 BUCH011576 BUCH011577 BUCH011578
Spider evaporators
Évaporateurs Évaporateurs rotatifs
Spider evaporators are ideal when users need to concentrate several samples under the same conditions.
  • Models for distillation with evaporating or cylindrical flasks available
  • Parallel distillation saves time