Imprimer
Erythrocyte sedimentation system, Sedi-Rate
Fournisseur: KARTELL
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
88435 88436 88437
KART88435EA 778 EUR
KART88435 KART88436 KART88437
Erythrocyte sedimentation system, Sedi-Rate
Systèmes de diagnostic clinique Prélèvement/préparation d'échantillons de diagnostic clinique Prélèvement/préparation d'échantillons de sang
Sedi-Rate allows a rapid and safe execution of the ESR test.
  • PP tube (lenght 86 mm) with label complete with pink fiber cap containing sodium citrate
  • Pipette in PS is silk-screened with a graduation scale
  • Piston allowing a quick filling of the pipette thanks to the pressure within the tube
According to the Westergren method.
Commander maintenant
SPÉCIFICATIONS
Environmentally Preferable