Imprimer
Stoppers for sample bottles, BEHROTEST®
Fournisseur: BEHR
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
B00168598 B00168597
719-1313EA 43.4 EUR
719-1313 719-1311
Stoppers for sample bottles, BEHROTEST®
Bouchons Bouchons en verre
These glass stoppers can be used instead of DIN/DEV diagonally cut stoppers. They are easy to use due to the lack of the easily broken edges.
  • No formation of bubbles below the stopper tip on inserting the stopper into the liquid sample
  • Less breakage or cracking risk than hollow stoppers