Imprimer
Pipettes monocanal, manuelles, Classic™
Fournisseur: RAININ
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
PR-10 PR-100 PR-1000 PR-10ML PR-20 PR-200 PR-5000 PR-START
RAINPR-10EA 621 EUR
RAINPR-10 RAINPR-100 RAINPR-1000 RAINPR-10ML RAINPR-20 RAINPR-200 RAINPR-5000 RAINPR-START
Pipettes monocanal, manuelles, Classic™
Pipettes Pipetteurs
Rainin Classic™ is a traditional, manual pipette with advanced ergonomic features, such as finger hook, light springs and silicone shock absorber.
  • Few out-of-tolerance errors
  • Rugged construction, stainless steel and quality PVDF for durability and easy maintenance
  • Ergonomic features ensure a hand-friendly operation and a reduced risk of injuries
  • Lower plunger forces: 25% less force required compared to similar traditional pipettes, thanks to lighter springs
  • Comfortable fingerhook for lighter grip and silicone shock absorber for less impact