Imprimer
RFID reader and LabX™ Direct Pipette-Scan™ software
Fournisseur: RAININ
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
RFID-KIT RFID-RDR
RAINRFID-KITEA 0 EUR
RAINRFID-KIT RAINRFID-RDR
RFID reader and LabX™ Direct Pipette-Scan™ software
Software Laboratory Software
RFID is "Radio Frequency IDentification" and with nothing more than Rainin's RFID reader and LabX™ Direct Pipette-Scan™ software, users can enhance how their lab manages pipettes. Rainin’s pipette management solution is easy to use and highly adaptable to different workflows. To be used with pipettes with RFID tag.
  • Software is compatible with Microsoft Windows XP/Vista/7
  • Once scanned, LabX displays the pipette’s complete profile, incl. serial number, manufacture and calibration due date, and any unique attributes assigned
  • Fast access to each pipette’s calibration and service data reduces the possibility of performance and/or compliance-related rework
  • LabX data easily exports to Microsoft Word and Excel, or as text for easy integration with other asset management applications
  • 12 customisable fields provide maximum flexibility
  • 'Read only' mode prevents pipette data from being overwritten
  • Service calibration data is always protected
Operation:
1. Scan a pipette's stored data (calibration and service, for example) by placing the pipette on the RFID reader
2. Create custom profiles for each pipette and transfer that data directly to the pipette’s RFID tag
3. Integrate your LabX data with third-party asset- and calibration-management applications (LabX data exports easily as Microsoft® Excel®, Word® or text files)