Imprimer
Immobilon® Blotting Filter Papers, Chromatography Grade
Fournisseur: Merck
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
IBFP0785C IBFP0813C
MILFIBFP0785CEA 126 EUR
MILFIBFP0785C MILFIBFP0813C
Immobilon® Blotting Filter Papers, Chromatography Grade
Papiers buvard
Chromotography-grade blotting paper for western and dot blotting.
  • Plain/white filter
  • Hydrophilic
  • Ambient shipping
For use with Amido black, CPTS, Colloidal gold, India ink and Ponceau-S red.
Chemiluminescent, colorimetric and fluorometric detection.