Imprimer
Value Plastics® Sanitary to Hose Barb Adapter, 1-1/2" Tri-Clamp to ³/₈" ID Hose Barb, ADCF PP; 100/PK
Fournisseur: Avantor Fluid Handling
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
VWR Item#MFLA31515-91BLK
Your Organization's Price:
Certificate of compliance included!
  • Animal derivative component free
  • Meet USP Class IV requirements
  • Sterilizable vis gamma irradiation
These sanitary fittings are the perfect to transition from sanitary clamp connections to seal or barb for flexible tubing. All adapters feature a radiused flow surface controlled to 15 micron Ra to ensure a smooth, uninterrupted flow—and no particulate build-up. There are no sharp edges in the flow path, so cleaning is very easy. Made from animal-derived ingredient-free polypropylene, these adapters offer good chemical resistance and may be sterilized by gamma irradiation. Material meets the requirements for USP Class VI.
<b>Informations de livraison: </b>Includes certificate of compliance.
En savoir plus
Other Products In this Family
See all Items in this Product Family