Imprimer
Masterflex® Quick-Disconnect Body, Acetal, 1/8" Flow, Valved, 1/4" Hose Barb
Fournisseur: Avantor Fluid Handling
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
VWR Item#MFLX70000-27
Your Organization's Price:
Unavailable
Disconnect and reconnect your flow path with ease.
  • Audible click lets you know the coupling is secure
  • Many hose barb sizes to choose from
  • Valved design allows you to cut off fluid flow when the fittings are disconnected
  • Material of construction is acetal│Has a 1/4" hose barb
Use these fittings to quickly connect and disconnect from your flow line. Simply order a body (female half) and an insert (male half). Couplings are compatible for use with gases and liquids. Choose between acetal for a wide operating temperature range and greater durability or polypropylene (PP) for stronger chemical resistance and the ability to be gamma sterilized.
En savoir plus
Other Products In this Family
See all Items in this Product Family