Imprimer
Blue-White® Variable-Area In-Line Flowmeter, Direct Read, Acrylic Housing, 1" NPT(F); 10 GPM Water
Fournisseur: Avantor Fluid Handling
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
VWR Item#MFLX32477-22
Your Organization's Price:
Read measurements in an unobstructed 360° view.
  • Easy-to-read dual scales (GPM/LPM)
  • Extremely durable—machined from high-quality acrylic rod stock
  • One-piece, in-line body occupies less space for a clean installation
  • Maximum flow rate of 10 GPM
These meters are designed for use in more aggressive process environments. In addition to the machined acrylic bodies, the design utilizes stainless steel float guides. The combination is a meter suitable for high-vibration, high-fluctuation process applications. Some versions include polypropylene end fittings reinforced with aluminum stress rings for added strength to the meters’ design.
En savoir plus
Other Products In this Family
See all Items in this Product Family