Imprimer
Filtres sans seringue, Mini-UniPrep™ G2, Whatman™
Fournisseur: Whatman products (Cytiva)
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
GN203APEORGSP GN203NPEORGSP GN203NPUORG GN203NPUORGSP GS203NPEORGSP GN203NPUGMFSP GN203APEORG GS503NPEORG
514-0386EA 556 EUR
514-0386 514-0391 514-0396 514-0397 514-0398 514-8140 514-8137 516-0480
Filtres sans seringue, Mini-UniPrep™ G2, Whatman™
Filtres Dispositifs de filtration sans seringue
Mini-UniPrep™ G2 is an 'all in one' syringeless filter with a borosilicate glass chamber. It replaces syringe filters for autosampler-supported UHPLC/HPLC sample preparation with easier handling and fewer consumable components.
  • Features an integrated autosampler vial, plunger with attached filter membrane, and septum/cap
  • Borosilicate glass vials eliminate plastic-based leachables that can originate from PP vials
  • Designed to hold up to 0,5 ml of unfiltered sample that can then be loaded directly into an autosampler
  • Easy to see when a sample has been
En savoir plus