Imprimer
Flacons YOUTILITY avec bouchon à vis, DURAN®
Fournisseur: DWK Life Sciences
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
218812854 218814452 218813653 218815457
215-4500EA 54.5 EUR
215-4500 215-4502 215-4501 215-4503
Flacons YOUTILITY avec bouchon à vis, DURAN®
Flacons Flacons en verre
DURAN®, borosilicate 3.3 glass, clear.
  • Specially shaped grip zones on the bottles for safer and easier handling
  • Ergonomic GL 45 screw cap made from food grade polypropylene
  • Shortened threads on bottle and cap for faster opening
  • Dedicated accessories for easy and simple identification and personalisation
  • Glass Type I according to USP, EP and JP
The DURAN® YOUTILITY bottle represents a new generation of glass laboratory bottles for use in a wide and diverse range of applications. The innovations include improved ergonomic bottle shape that extends toEn savoir plus