Imprimer

High Power LED ring light (HPRL), Segmentable (D)

Fournisseur: ZEISS
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : sur 5

4170409300000
ZEIS4170409300000EA 0 EUR
ZEIS4170409300000
High Power LED ring light (HPRL), Segmentable (D)
Éclairages Éclairage LED
This product is designed for lighting tasks in the field of stereo microscopy for industrial and developed and intended for laboratory use. A lens focuses the light at the desired working distance.

  • 8 high power LEDs with ring optics, optics mountable for close or long range
  • Color temperature approx. 5800 K, lifespan approx. 25000 h (lumen maintenance 70%)
  • Ergonomic operation on the housing: No separate controller required
  • Rotary encoder 1 for setting segment modes, rotation in 45° steps and autorotation
  • Rotary encoder 2 for dimming, saving and recalling a
En savoir plus
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR