Imprimer

pH Combination Electrode

Fournisseur: WTW
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : sur 5

EGA173I
WTWAEGA173IEA 225 EUR
WTWAEGA173I
pH Combination Electrode
Électrodes Électrodes pH
pH Combination electrode for heavily polluted waste water and electroplating.

  • Ring-shaped sleeve junction
  • Gel electrolyte with KCl reserve pipe

Application are drinking water, water soluble lacquers, low ionic mediums, electroplating, waste water.

Membrane shape: Cylinder
Isopotential point: 7,0 ±0,3
Pressure: <6 bar
Minimum conductivity of the medium: 50 µS/cm
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR