Imprimer
Culture flasks with rim, BIOGEN
Fournisseur: witeg Labortechnik
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : sur 5
5.507.204 5.507.201 5.507.203 5.507.205
WITG5.507.204EA 62 EUR
WITG5.507.204 WITG5.507.201 WITG5.507.203 WITG5.507.205
Culture flasks with rim, BIOGEN
Ballons Flacons de culture
Culture flask, made of borosilicate glass, Erlenmeyer form, designed for orbital shakers.
  • Neck with rim
  • Triple baffled shake flask
  • Excellent chemical and temperature resistance