Imprimer
Culture flasks, Erlenmeyer shape, BIOGEN
Fournisseur: witeg Labortechnik
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : sur 5
5.507.108 5.507.106 5.507.104 5.507.107 5.507.102
WITG5.507.108EA 109 EUR
WITG5.507.108 WITG5.507.106 WITG5.507.104 WITG5.507.107 WITG5.507.102
Culture flasks, Erlenmeyer shape, BIOGEN
Ballons Flacons de culture
Culture flasks, made of borosilicate glass, Erlenmeyer shape, designed for orbital shakers.
  • Straight neck for metal caps
  • Triple baffled shake flask
  • Excellent chemical and temperature resistance