Imprimer
Hybridisation bottles
Fournisseur: witeg Labortechnik
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : sur 5
2.446.115
WITG2.446.115EA 59 EUR
WITG2.446.115
Hybridisation bottles
Flacons Flacons d'hybridation
Borosilicate glass 3.3, with GL 45 screw cap and silicone sealing.
  • Excellent chemical resistance
  • High temperature resistance
  • Minimal thermal expansion giving relatively high resistance to temperature changes