Imprimer
Drying tubes, U-shaped
Fournisseur: witeg Labortechnik
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : sur 5
2.114.100 2.114.125 2.114.150 2.114.250
WITG2.114.100EA 69 EUR
WITG2.114.100 WITG2.114.125 WITG2.114.150 WITG2.114.250
Drying tubes, U-shaped
Tubes Tubes de dessication
Borosilicate glass, clear. Replaceable NS stopcocks. Lateral tube connectors.
  • Very good chemical resistance
  • High temperature resistance
  • Minimal thermal expansion giving relatively high resistance to temperature changes