Imprimer
Entonnoirs à joint conique
Fournisseur: witeg Labortechnik
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : sur 5
1.650.014 1.650.029 1.650.129 1.650.130 1.650.150 1.650.019
WITG1.650.014EA 13 EUR
WITG1.650.014 WITG1.650.029 WITG1.650.129 WITG1.650.130 WITG1.650.150 WITG1.650.019
Entonnoirs à joint conique
Entonnoirs Entonnoirs pour la filtration Entonnoirs à joints coniques
Funnels, made of boroslicate glass, with standard ground joint cone.
  • Very good chemical resistance
  • High temperature resistance
  • Minimal thermal expansion giving relatively high resistance to temperature changes
Ideal for transferring liquids or powders to glassware with standard ground joints.