Imprimer
Attachable screw thread tubes
Fournisseur: witeg Labortechnik
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : sur 5
0.182.315
WITG0.182.315EA 2.7 EUR
WITG0.182.315
Attachable screw thread tubes
Adaptateurs Adaptateurs pour tubes filetés
DURAN®, borosilicate glass 3.3, clear.
  • Excellent chemical resistance
  • High temperature resistance
  • Minimal thermal expansion giving relatively high resistance to temperature changes
DIN screw thread SVL.