Imprimer
Micro spatula for NMR sample tubes, SP Wilmad-LabGlass
Fournisseur: SP Industries
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : sur 5
806
WILM806EA 63.8 EUR
WILM806
Micro spatula for NMR sample tubes, SP Wilmad-LabGlass
Spatules
Stainless steel. This spatula fits into the most common NMR tubes. It enables users to get a tiny powder sample into any tube larger than 4 mm ext. Ø.
Ideal for adding or removing samples (including freeze-dried). Scoop samples with the round end or pry stiff samples with the sharp-edged flat end.