Imprimer
Drum opener, WHEATON®
Fournisseur: DWK Life Sciences
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : sur 5
885295
WHEA885295EA 92.7 EUR
WHEA885295
Drum opener, WHEATON®
Clés à molette
These drum openers can be used with hazardous materials. Made from lightweight, durable aluminum alloy.
  • Quarter-turn opening action
Commander maintenant
SPÉCIFICATIONS
Environmentally Preferable