Imprimer
Complete sampler, Wheaton® E-Z
Fournisseur: DWK Life Sciences
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : sur 5
885020
WHEA885020EA 359 EUR
WHEA885020
Complete sampler, Wheaton® E-Z
Échantillonneurs Préleveurs de liquides
The filling ports are located above the bottle to ensure that it is completely filled to reduce the possibility of air entrapment.
  • Head/shaft assembly and borosilicate glass bottle with lined cap
  • Constructed of chemically resistant PP
  • Bottle with a GPI 33-430 thread
This sampler is an efficient and time saving method of sampling from drums and tanks.