Imprimer
Vacuum filtration complete units, WHEATON®
Fournisseur: DWK Life Sciences
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : sur 5
419380 419385-47
WHEA419380EA 557 EUR
WHEA419380 WHEA419385-47
Vacuum filtration complete units, WHEATON®
Entonnoirs Entonnoirs filtrants avec membrane
Filtration assemblies with tapered joints, metal clamp and support base.
  • Recommended for general filtration
  • Units have a hose connection for ¼" (6 mm) Int.Ø flexible tubing
Recommended for routine filtration of organic solvents, corrosive liquids and the removal of particulates from HPLC solvents. Ground glass connection eliminates phthalate contamination that can occur from silicone or neoprene stoppers. Drip tip on the support bases extends below the hose connection preventing filtrate from entering the vacuum line.
Glass components manufacturedEn savoir plus