Imprimer
Grade 0903 filter papers for technical use, Whatman™
Fournisseur: Whatman products (Cytiva)
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : sur 5
10334885
WHAT10334885EA 496 EUR
WHAT10334885
Grade 0903 filter papers for technical use, Whatman™
Filtres Papiers filtre Papiers filtres qualitatifs
Thin filter paper with smooth surface. Offers medium to slow flow rate and good retention for small particles.
These filter papers are made from super-refined cellulose and have been specifically designed to have particular properties for each technical application. The applications range from the filtration of beverages to the purification of electroplating baths.