Imprimer
VWR® Gas Venting Filters
Fournisseur: VWR Collection
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : sur 5
VWRI210-000121EA 520 EUR
VWRI210-000121 VWRI210-000125 VWRI210-000129 VWRI210-000122 VWRI210-000126 VWRI210-000123 VWRI210-000127 VWRI210-000124 VWRI210-000128
VWR® Gas Venting Filters
Filtres Filtres de ventilation
VWR® Gas venting filters are designed for obstructing bacterial, particles or moisture when venting in different applications. Sterile venting of bioreactors, fermentation tanks, media flasks, and carboys. Sterile gas purge of cell culture vessels or filling vessels. In-line sterilisation of and particulate removal from air and gases. Obstruct moisture to protect equipment.
  • Sterile packed filters are ready to use
  • 100% class VI resin
  • Optimised hydrophobic PTFE membrane provides superior flow rates in a compact device
  • Integrity tested with bubble point
En savoir plus