Imprimer
Dust cloths, standard and high performance
Fournisseur: FHP VILEDA SCS
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : sur 5
133909
VILE133909EA 17.6 EUR
VILE133909
Dust cloths, standard and high performance
Lingettes Chiffons nettoyants réutilisables Chiffons pour applications générales
These dust-binding cloths are impregnated with a care oil.
  • Pre-impregnated surface collects dust like a magnet
  • Products retain their effectiveness even after being stored for some time
  • High Performance cloth reduces number of airborne particles
The Standard cloth is ideal for the dry cleaning of hard floor surfaces. The High Performance cloth is ideal for the dry cleaning of hospitals, offices and computer rooms.