Imprimer
Cleanroom wipes, pre-saturated, DAS-WIPE 100 Sterile, VAI®
Fournisseur: VELTEK ASSOCIATES
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : sur 5
DW1001212S2319
VELTDW1001212S2319EA 1280 EUR
VELTDW1001212S2319
Cleanroom wipes, pre-saturated, DAS-WIPE 100 Sterile, VAI®
Lingettes Lingettes pour salles propres Lingettes pour salles propres, présaturées
Individually-packaged sterile wipes are a polyester blend that is inherently low in particulate and shedding features and saturated with DAS Stainless Steel Cleaner and Lubricant. DAS-Wipe 100 is designed to replace silicone on turntables, process lines and as a general lubricant to assure bottles do not stick during movement towards the critical fill site.
  • Laundered and packaged in ISO 4 conditions, closed filling in an ISO 5 Cleanroom (Grade A)
  • 100% synthetic filament constructed in a continuous knit that minimizes fiber and particulate release
En savoir plus