Imprimer

Anti-Peptide Histidine-Methionine Rabbit Polyclonal Antibody

Fournisseur: US Biological
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : sur 5

P3282-05A
USBIP3282-05AEA 561 EUR
USBIP3282-05A
Anti-Peptide Histidine-Methionine Rabbit Polyclonal Antibody
Anticorps
Type: Primary
Antigen: Peptide Histidine-Methionine
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR