Imprimer

Anti-Macrophage Colony Stimulating Factor Rabbit Polyclonal Antibody (Biotin)

Fournisseur: US Biological
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : sur 5

M1201-02C-BIOT
USBIM1201-02C-BIOTEA 884 EUR
USBIM1201-02C-BIOT
Anti-Macrophage Colony Stimulating Factor Rabbit Polyclonal Antibody (Biotin)
Anticorps
Type: Primary
Antigen: Macrophage Colony Stimulating Factor
Clonality: polyclonal
Clone:
Conjugation: Biotin
Epitope: C-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR