Imprimer
Régulateurs de contre-pression, Upchurch Scientific®
Fournisseur: UPCHURCH SCIENTIFIC
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : sur 5
P-761 P-762 P-763 P-764 P-765 P-785 P-786 P-787 P-790 P-791 P-795 P-796 P-880 P-455 U-605 U-606 U-607 U-608 U-609 U-610
UPCHP-761EA 217 EUR
UPCHP-761 UPCHP-762 554-3182 UPCHP-764 UPCHP-765 554-3197 554-3198 554-3199 UPCHP-790 554-3200 UPCHP-795 UPCHP-796 554-3203 554-3116 UPCHU-605 UPCHU-606 554-3270 UPCHU-608 UPCHU-609 UPCHU-610
Régulateurs de contre-pression, Upchurch Scientific®
Vannes Clapets anti-retour
Back pressure regulator assemblies are designed to create incremental back pressures ranging from 5 to 1000 psi (0,3 to 69 bar). These biocompatible and stainless steel back pressure regulator assemblies are available complete with a replaceable, factory pre-set cartridge (except the 5 and 20 psi (0,3 to 1,4 bar versions).
The back pressure regulator cartridges are manufactured from PEEK, ETFE, perfluoroelastomer, and gold-plated stainless steel giving it excellent chemical resistance. They will easily operate in any of the standard back pressure regulator holders. These cartridges createEn savoir plus