Imprimer
Cleaning Swabs for Fiber Optic Sleeve (2,5 mm)
Fournisseur: Texwipe
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : sur 5
48040
TEXW48040EA 474 EUR
TEXW48040
Cleaning Swabs for Fiber Optic Sleeve (2,5 mm)
Écouvillons Écouvillons de nettoyage
These cleaning swabs are versatile, effective, and cost-efficient swab for cleaning end faces and equipment test ports.
  • 2,5 mm flexible swab head conforms to connector face
  • No adhesives, binders, or fibers to leave residues
  • Can be used with all Electro-Wash® and Fiber-Wash™ Precision fiber optic cleaners for all types of soils and contaminants
  • 2,5 mm thick head
Applications:
- Cleans 2,5 mm connectors and ferrules
- Cleans test equipment ports