Imprimer
delta P air flow ComboKit 1 with Bluetooth®, Testo 440
Fournisseur: TESTO
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : sur 5
0563.4409
TEST0563.4409EA 1820 EUR
TEST0563.4409
delta P air flow ComboKit 1 with Bluetooth®, Testo 440
Enregistreurs de données
Delta P air flow ComboKit 1 equipped with clearly structured measurement menu for volume flow and parallel determination of air velocity, differential pressure, humidity and temperature in ventilation ducts or at ventilation outlets.
  • Carry out measurements without any annoying cables
  • With space-saving and integrated differential pressure sensor
  • Easily transported
Convenient with wireless probes, internal data storage and USB port for data export. Ideal for long-term monitoring of indoor air quality and maximum flexibility for duct and outletEn savoir plus