Imprimer
Hanging racks
Fournisseur: TEKNOMEK
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : sur 5
CRGS-HR2H WR0206/316 WR0303 WR0319 WR0323 CRGS-HR8H CRGS-HR12H WR0315
TEKMCRGS-HR2HEA 0 EUR
TEKMCRGS-HR2H TEKMWR0206/316 TEKMWR0303 TEKMWR0319 TEKMWR0323 TEKMCRGS-HR8H TEKMCRGS-HR12H TEKMWR0315
Hanging racks
Mobilier Portemanteaux
Our stainless-steel hanging racks are manufactured to the highest standards.
  • Brushed dull polish finish
  • Wall mounted
  • Optional number etching if required
Our hanging racks are manufactured with 304 grade stainless steel for optimised durability and strength.
Long-lasting and sturdy, they are designed to the highest standard, ensuring super-smooth edges to minimise scratching and snagging.
Wall mounted, enabling space-efficiency. As with all our products, our stainless-steel coat racks are suited to rigorous hygiene and HACCP routines, meaning youEn savoir plus